3-Fluoro-4-(methoxycarbamoyl)phenylboronic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: (3-Fluoro-4-(methoxycarbamoyl)phenyl)boronic acid, 3-Fluoro-4-(methoxycarbamoyl)benzeneboronic acid

H52610.MD H52610.03
H52610.MDEA 0 EUR
H52610.MD H52610.03
3-Fluoro-4-(methoxycarbamoyl)phenylboronic acid ≥97%
3-Fluoro-4-(methoxycarbamoyl)phenylboronic acid

Purity: 97% CAS No.: 913835-58-2 Molecular Formula: C8H9BFNO4 Formula Weight: 212.97 Melting Point: 161-163° MDL No.: MFCD08436059

Formula: C₈H₉BFNO₄
MW: 212,97 g/mol
Melting Pt: 161…163 °C
Storage Temperature: Ambient
MDL Number: MFCD08436059
CAS Number: 913835-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR