2-(Trifluoromethoxy)thiobenzamide ≥97%

Supplier: Thermo Scientific
Danger

H33047.03 H33047.06
H33047.03EA 83.4 EUR
H33047.03 H33047.06
2-(Trifluoromethoxy)thiobenzamide ≥97%
2-(Trifluoromethoxy)thiobenzamide

Purity: 97% CAS No.: 926247-77-0 Molecular Formula: C8H6F3NOS Formula Weight: 221.20 Melting Point: 82-86° MDL No.: MFCD09042874

Formula: C₈H₆F₃NOS
MW: 221,20 g/mol
Melting Pt: 82…86 °C
Storage Temperature: Ambient
MDL Number: MFCD09042874
CAS Number: 926247-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR