3-(Benzyloxy)phenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: (3-(Benzyloxy)phenyl)boronic acid, 3-Benzyloxybenzeneboronic acid

L17474.03 L17474.06
L17474.03EA 0 EUR
L17474.03 L17474.06
3-(Benzyloxy)phenylboronic acid ≥98%
3-(Benzyloxy)phenylboronic acid

Purity: 98+% CAS No.: 156682-54-1 Molecular Formula: C13H13BO3 Formula Weight: 228.06 Melting Point: 122-124° Beilstein Reference: 6808244

Formula: C₁₃H₁₃BO₃
MW: 228,06 g/mol
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD02093063
CAS Number: 156682-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR