6-Quinolineboronic acid hydrochloride 95%

Supplier: Thermo Scientific
Warning

H30380.MD H30380.03
H30380.MDEA 0 EUR
H30380.MD H30380.03
6-Quinolineboronic acid hydrochloride 95%
6-Quinolineboronic acid hydrochloride

Purity: 95% Molecular Formula: C9H8BNO2.HCl Formula Weight: 209.44 Melting Point: ca 220° dec. MDL No.: MFCD11101398

Formula: C₉H₉BClNO₂
MW: 209,44 g/mol
Melting Pt: ∼ 220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD11101398
CAS Number: 1310403-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR