Isoquinoline-1-carbonitrile ≥99%

Supplier: Thermo Scientific
Warning

Synonyms: 1-Isoquinolinecarbonitrile

L19425.03 L19425.06
L19425.03EA 64.7 EUR
L19425.03 L19425.06
Isoquinoline-1-carbonitrile ≥99%
Isoquinoline-1-carbonitrile

Purity: 99% CAS No.: 1198-30-7 Molecular Formula: C10H6N2 Formula Weight: 154.17 Odour: Odourless Melting Point: 87-91° Beilstein Reference: 119104 MDL No.: MFCD00134166

Formula: C₁₀H₆N₂
MW: 154,17 g/mol
Melting Pt: 87…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00134166
CAS Number: 1198-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR