5-Methyl-3-(4-methylphenyl)-1,2,4-oxadiazole ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
H54399.MD
H54399.03
H54399.MDEA
0
EUR
H54399.MD
H54399.03
5-Methyl-3-(4-methylphenyl)-1,2,4-oxadiazole ≥97%
5-Methyl-3-(4-methylphenyl)-1,2,4-oxadiazole
Purity: 97%
CAS No.: 81386-30-3
Molecular Formula: C10H10N2O
Formula Weight: 174.20
Beilstein Reference: 7438
MDL No.: MFCD00085801
Formula:
C₁₀H₁₀N₂O Storage Temperature: Ambient |
MDL Number:
MFCD00085801 CAS Number: 81386-30-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...