3,5-Dichloropyridine-2-carbonitrile 95%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
3,5-Dichloropicolinonitrile, 3,5-Dichloro-2-cyanopyridine
L19723.06
L19723.14
L19723.06EA
0
EUR
L19723.06
L19723.14
3,5-Dichloropyridine-2-carbonitrile 95%
3,5-Dichloropyridine-2-carbonitrile
Purity: 95%
CAS No.: 85331-33-5
Molecular Formula: C6H2Cl2N2
Formula Weight: 173.00
Melting Point: 99-103°
Beilstein Reference: 4390101
UN No.: 3439
MDL No.: MFCD03788758
Formula:
C₆H₂Cl₂N₂ MW: 173,00 g/mol Melting Pt: 99…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788758 CAS Number: 85331-33-5 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...