5-(3-Bromophenyl)-1H-tetrazole ≥97%
Supplier: Thermo Scientific
|
Warning
|
L15180.03
L15180.MD
L15180.03EA
0
EUR
L15180.03
L15180.MD
5-(3-Bromophenyl)-1H-tetrazole ≥97%
5-(3-Bromophenyl)-1H-tetrazole
Purity: 97%
CAS No.: 3440-99-1
Molecular Formula: C7H5BrN4
Formula Weight: 225.05
Melting Point: 150-152°
Beilstein Reference: 141258
Formula:
C₇H₅BrN₄ MW: 225,05 g/mol Melting Pt: 150…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075675 CAS Number: 3440-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...