3-Aminopropionitrile 98% stabilised
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
β-Aminopropionitrile, 3-Aminopropanenitrile
351750050
351750010
351750250
ACRO351750050EA
121
EUR
ACRO351750050
ACRO351750010
ACRO351750250
3-Aminopropionitrile 98% stabilised
3-Aminopropionitrile
Formula:
C₃H₆N₂ MW: 70,10 g/mol Boiling Pt: 185 °C (760 mmHg) Density: 0,9584 g/cm³ Flash Pt: 85 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00014820 CAS Number: 151-18-8 EINECS: 205-786-0 UN: 2922 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4365 to 1.4395 (20°C, 589 nm) |
GC | ≥97.5 % |
Stabilizer | potassium carbonate |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...