Ethyl trifluoropyruvate ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Ethyl 3,3,3-trifluoro-2-oxopropanoate, Ethyl 3,3,3-trifluoropyruvate
B20611.06
B20611.14
B20611.06EA
65.1
EUR
B20611.06
B20611.14
Ethyl trifluoropyruvate ≥97%
Ethyl trifluoropyruvate
Purity: 97%
CAS No.: 13081-18-0
Molecular Formula: C5H5F3O3
Formula Weight: 170.09
Flash Point: 27°(80°F)
Refractive Index: 1.3405
Boiling Point: 95-98°
Beilstein Reference: 2087388
UN No.: 3272
MDL No.: MFCD00114935
Formula:
C₅H₅F₃O₃ MW: 170,09 g/mol Boiling Pt: 95…98 °C Density: 1,400 Flash Pt: 27 °C (80 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00114935 CAS Number: 13081-18-0 EINECS: 000-000-0 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...