3,4-Difluorothiobenzamide ≥97%

Supplier: Thermo Fisher Scientific
Warning

H54564.MD H54564.03
H54564.MDEA 0 EUR
H54564.MD H54564.03
3,4-Difluorothiobenzamide ≥97%
3,4-Difluorothiobenzamide

Purity: 97% CAS No.: 317319-14-5 Molecular Formula: C7H5F2NS Formula Weight: 173.18 MDL No.: MFCD04036083

Formula: C₇H₅F₂NS
Storage Temperature: Ambient
MDL Number: MFCD04036083
CAS Number: 317319-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR