4-Butoxybenzonitrile ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
A16570.06
A16570.14
A16570.06EA
56.9
EUR
A16570.06
A16570.14
4-Butoxybenzonitrile ≥97%
4-Butoxybenzonitrile
Purity: 97%
CAS No.: 5203-14-5
Molecular Formula: C11H13NO
Formula Weight: 175.23
Flash Point:
Formula:
C₁₁H₁₃NO Boiling Pt: 242…244 °C Density: 0,999 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00043482 CAS Number: 5203-14-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...