5-Isoquinolinecarboxylic acid 96%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Isoquinoline-5-carboxylic acid
L19637.03
L19637.MD
L19637.03EA
0
EUR
L19637.03
L19637.MD
5-Isoquinolinecarboxylic acid 96%
5-Isoquinolinecarboxylic acid
Purity: 96%
CAS No.: 27810-64-6
Molecular Formula: C10H7NO2
Formula Weight: 173.17
Melting Point: 278-281°
Boiling Point: 60-62°/11mm
MDL No.: MFCD03788744
Formula:
C₁₀H₇NO₂ MW: 173,17 g/mol Boiling Pt: 60…62 °C (11 mmHg) Melting Pt: 278…281 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788744 CAS Number: 27810-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...