(1R,3S)-(+)-Camphoric acid ≥98%
Supplier: Thermo Scientific
Synonyms:
D(+)-Camphoric acid, (1R,3S)-1,2,2-Trimethylcyclopentane-1,3-dicarboxylic acid, Levocarnitine Impurity B
B23106.14
B23106.22
B23106.36
B23106.14EA
34.5
EUR
B23106.14
B23106.22
B23106.36
(1R,3S)-(+)-Camphoric acid ≥98%
(1R,3S)-(+)-Camphoric acid
Purity: 98+%
CAS No.: 124-83-4
Molecular Formula: C10H16O4
Formula Weight: 200.23
Melting Point: 183-188°
Beilstein Reference: 2050204
Merck Reference: 14,1733
Optical Rotation: +46° (c=1 in methanol)
MDL No.: MFCD00001375
Formula:
C₁₀H₁₆O₄ MW: 200,23 g/mol Melting Pt: 183…188 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001375 CAS Number: 124-83-4 EINECS: 204-715-0 Merck Index: 14,1733 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...