Ethyl-4-chloro-6-fluoroquinoline-3-carboxylate 95%

Supplier: Thermo Scientific
Warning

Synonyms: Ethyl 4-chloro-6-fluoroquinoline-3-carboxylate

H50502.03 H50502.MD
H50502.03EA 0 EUR
H50502.03 H50502.MD
Ethyl-4-chloro-6-fluoroquinoline-3-carboxylate 95%
Ethyl 4-chloro-6-fluoroquinoline-3-carboxylate

Purity: 95% CAS No.: 77779-49-8 Molecular Formula: C12H9ClFNO2 Formula Weight: 253.66 MDL No.: MFCD00173350

Formula: C₁₂H₉ClFNO₂
MW: 253,66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173350
CAS Number: 77779-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR