Ethyl-4-chloro-8-fluoroquinoline-3-carboxylate ≥97%

Supplier: Thermo Scientific

Synonyms: Ethyl 4-chloro-8-fluoroquinoline-3-carboxylate

H50507.03 H50507.MD
H50507.03EA 351 EUR
H50507.03 H50507.MD
Ethyl-4-chloro-8-fluoroquinoline-3-carboxylate ≥97%
Ethyl 4-chloro-8-fluoroquinoline-3-carboxylate

Purity: 97% CAS No.: 56824-90-9 Molecular Formula: C12H9ClFNO2 Formula Weight: 253.66 MDL No.: MFCD00173364

Formula: C₁₂H₉ClFNO₂
MW: 253,66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173364
CAS Number: 56824-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR