Ethyl-4-chloro-8-methylquinoline-3-carboxylate

Supplier: Thermo Fisher Scientific

H50510.03 H50510.MD
H50510.03EA 0 EUR
H50510.03 H50510.MD
Ethyl-4-chloro-8-methylquinoline-3-carboxylate
Ethyl 4-chloro-8-methylquinoline-3-carboxylate

CAS No.: 37041-32-0 Molecular Formula: C13H12ClNO2 Formula Weight: 249.70 MDL No.: MFCD00173399

Formula: C₁₃H₁₂ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD00173399
CAS Number: 37041-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR