4-(3-Methylphenoxy)piperidine ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-(m-Tolyloxy)piperidine
H50950.03
H50950.MD
H50950.03EA
154
EUR
H50950.03
H50950.MD
4-(3-Methylphenoxy)piperidine ≥97%
4-(3-Methylphenoxy)piperidine
Purity: 97%
CAS No.: 63843-46-9
Molecular Formula: C12H17NO
Formula Weight: 191.28
Melting Point: 190-191°
MDL No.: MFCD06248345
Formula:
C₁₂H₁₇NO MW: 191,28 g/mol Melting Pt: 190…191 °C Storage Temperature: Ambient |
MDL Number:
MFCD06248345 CAS Number: 63843-46-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...