Direct red 80

Supplier: Thermo Fisher Scientific

Synonyms: CI 35780, Sirius Red F3B, Sirius Red, Picro-Sirius Red

B21693.06 B21693.14 B21693.22
B21693.06EA 35.3 EUR
B21693.06 B21693.14 B21693.22
Direct red 80
Direct Red 80

CAS No.: 2610-10-8 Molecular Formula: C45H26N10Na6O21S6 Formula Weight: 1373.09 MDL No.: MFCD00054389

Formula: C₄₅H₂₆N₁₀Na₆O₂₁S₆
Storage Temperature: Ambient
MDL Number: MFCD00054389
CAS Number: 2610-10-8
EINECS: 220-027-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR