2-(1-Naphthalene)ethanol 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-(Naphthalen-1-yl)ethanol, 2-(1-Naphthyl)ethanol
H27218.09
H27218.09EA
79.8
EUR
H27218.09
2-(1-Naphthalene)ethanol 95%
2-(1-Naphthalene)ethanol
Purity: 95%
CAS No.: 773-99-9
Molecular Formula: C12H12O
Formula Weight: 172.23
Melting Point: 60-64°
Boiling Point: 186°/17mm
MDL No.: MFCD00004050
Formula:
C₁₂H₁₂O MW: 172,23 g/mol Boiling Pt: 186 °C (17 mmHg) Melting Pt: 60…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004050 CAS Number: 773-99-9 EINECS: 212-260-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...