4-Bromo-4'-methylbiphenyl ≥97%
Supplier: Thermo Scientific
Synonyms:
4-Bromo-4'-methyl-1,1'-biphenyl
H50596.03
H50596.03EA
451
EUR
H50596.03
4-Bromo-4'-methylbiphenyl ≥97%
4-Bromo-4'-methylbiphenyl
Purity: 97%
CAS No.: 50670-49-0
Molecular Formula: C13H11Br
Formula Weight: 247.14
Melting Point: 136°
MDL No.: MFCD00466236
Formula:
C₁₃H₁₁Br MW: 247,14 g/mol Melting Pt: 136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00466236 CAS Number: 50670-49-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...