Trimesic acid triethyl ester ≥97%
Supplier: Thermo Scientific
Synonyms:
Triethyl 1,3,5-benzenetricarboxylate, Triethyl benzene-1,3,5-tricarboxylate
B23392.06
B23392.14
B23392.06EA
39.4
EUR
B23392.06
B23392.14
Trimesic acid triethyl ester ≥97%
Trimesic acid triethyl ester
Purity: 97%
CAS No.: 4105-92-4
Molecular Formula: C15H18O6
Formula Weight: 294.31
Melting Point: 132-135°
MDL No.: MFCD00236389
Formula:
C₁₅H₁₈O₆ MW: 294,31 g/mol Melting Pt: 132…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236389 CAS Number: 4105-92-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...