HPLC columns, Acclaim™ Phenyl-1
Supplier: Thermo Fisher Scientific
071969 071970 071971 071973 071974 079697
555-7171EA 0 EUR
555-7171 555-7172 555-7173 555-7175 555-7176 555-7528
HPLC columns, Acclaim™ Phenyl-1
Chromatography Columns
These columns are widely used in pharmaceutical, environmental, food testing and product quality testing and it is suitable for the analysis of the aromatic analytes.
  • High aromatic selectivity
  • High hydrophobic retention
  • Compatibility with high aqueous mobile phase
  • High efficiency and rugged packing