2,2'-Bithiophene-5-boronic acid pinacol ester ≥98%

Supplier: Thermo Scientific

Synonyms: 2-([2,2'-Bithiophen]-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H37989.03 H37989.06
H37989.03EA 46.4 EUR
H37989.03 H37989.06
2,2'-Bithiophene-5-boronic acid pinacol ester ≥98%
2,2'-Bithiophene-5-boronic acid pinacol ester
Formula: C₁₄H₁₇BO₂S₂
MW: 292,2 g/mol
Melting Pt: 35…38 °C
Storage Temperature: Freezer
MDL Number: MFCD04039974
CAS Number: 479719-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR