Ethyl-2-aminoisonicotinate 97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Amino-isonicotinic acid ethyl ester, Ethyl 2-aminopyridine-4-carboxylate, 2-Aminopyridine-4-carboxylic acid Ethyl Ester, Ethyl 2-aminoisonicotinate, Ethyl 2-Aminopyridine-4-carboxylate, 2-Aminoisonicotinic acid Ethyl Ester, Ethyl 2-Aminoisonicotinate

H61280.06 H61280.14
H61280.06EA 153 EUR
H61280.06 H61280.14
Ethyl-2-aminoisonicotinate 97%
Ethyl 2-aminoisonicotinate
Formula: C₈H₁₀N₂O₂
MW: 166,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03791260
CAS Number: 13362-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR