Zirconium diboride ≥99.5% (metals basis excluding Hf)
Supplier: Thermo Fisher Scientific
|
Warning
|
47030.18
47030.30
47030.18EA
148
EUR
47030.18
47030.30
Zirconium diboride ≥99.5% (metals basis excluding Hf)
Zirconium diboride
Formula:
B₂Zr Melting Pt: 3000 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064648 CAS Number: 12045-64-6 EINECS: 234-963-5 UN: 3178 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...