α,4-Dibromo-3-fluorotoluene 96%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-Bromo-3-fluorobenzyl bromide, 1-Bromo-4-(bromomethyl)-2-fluorobenzene

H59275.MD H59275.03
H59275.MDEA 0 EUR
H59275.MD H59275.03
α,4-Dibromo-3-fluorotoluene 96%
α,4-Dibromo-3-fluorotoluene
Formula: C₇H₅Br₂F
Melting Pt: 33…36 °C
Storage Temperature: Ambient
MDL Number: MFCD11847179
CAS Number: 127425-73-4
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR