4-Fluoro-4'-methoxybenzophenone ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(4-Fluorophenyl)(4-methoxyphenyl)methanone
H61679.MD
H61679.06
H61679.MDEA
0
EUR
H61679.MD
H61679.06
4-Fluoro-4'-methoxybenzophenone ≥98%
4-Fluoro-4'-methoxybenzophenone
Formula:
C₁₄H₁₁FO₂ MW: 230,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00055469 CAS Number: 345-89-1 EINECS: 206-464-2 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...