4-Cyano-2-fluorophenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Cyano-2-fluorophenylboronic Acid, 4-Cyano-2-fluorobenzeneboronic acid

H61134.MD H61134.03
H61134.MDEA 0 EUR
H61134.MD H61134.03
4-Cyano-2-fluorophenylboronic acid ≥98%
4-Cyano-2-fluorophenylboronic acid

Purity: 98% CAS No.: 1150114-77-4 Molecular Formula: C7H5BFNO2 Formula Weight: 164.93 Physical Form: Solid

Formula: C₇H₅BFNO₂
MW: 164,93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07644472
CAS Number: 1150114-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR