4-Amino-2-bromobenzonitrile 95%
Supplier: Thermo Scientific
|
Warning
|
H61247.03
H61247.06
H61247.03EA
82.8
EUR
H61247.03
H61247.06
4-Amino-2-bromobenzonitrile 95%
4-Amino-2-bromobenzonitrile
Purity: 95%
CAS No.: 53312-82-6
Molecular Formula: C7H5BrN2
Formula Weight: 197.03
Physical Form: Powder
Appearance: Yellow
UN No.: 3439
MDL No.: MFCD03695814
Formula:
C₇H₅BrN₂ MW: 197,03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03695814 CAS Number: 53312-82-6 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...