2-Bromo-p-anisaldehyde ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Bromo-4-methoxybenzaldehyde

H61379.MD H61379.03 H61379.06
H61379.MDEA 0 EUR
H61379.MD H61379.03 H61379.06
2-Bromo-p-anisaldehyde ≥98%
2-Bromo-p-anisaldehyde
Formula: C₈H₇BrO₂
MW: 215.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09788636
CAS Number: 43192-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR