Methyl-3-bromo-2-fluorobenzoate ≥98%
Supplier: Thermo Fisher Scientific
Synonyms:
Methyl 3-bromo-2-fluorobenzoate
H36865.03
H36865.06
H36865.03EA
0
EUR
H36865.03
H36865.06
Methyl-3-bromo-2-fluorobenzoate ≥98%
Methyl 3-bromo-2-fluorobenzoate
Formula:
C₈H₆BrFO₂ Melting Pt: 35…37 °C Storage Temperature: Ambient |
MDL Number:
MFCD09261255 CAS Number: 206551-41-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...