Methyl-6-(hydroxymethyl)picolinate 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Methyl 6-(hydroxymethyl)picolinate, Methyl 6-(hydroxymethyl)pyridine-2-carboxylate

H63793.MD H63793.03
H63793.MDEA 0 EUR
H63793.MD H63793.03
Methyl-6-(hydroxymethyl)picolinate 95%
Methyl 6-(hydroxymethyl)picolinate

Purity: 95% CAS No.: 39977-44-1 Molecular Formula: C8H9NO3 Formula Weight: 167.16 Physical Form: Solid Appearance: White MDL No.: MFCD03840850

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03840850
CAS Number: 39977-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR