Bis(cyclopentadienyl)hafnium dichloride
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Hafnocene dichloride
Packaged under argon.
36105.03
36105.06
36105.14
36105.03EA
57.3
EUR
36105.03
36105.06
36105.14
Bis(cyclopentadienyl)hafnium dichloride
Bis(cyclopentadienyl)hafnium dichloride
CAS No.: 12116-66-4
Molecular Formula: C10H10Cl2Hf
Formula Weight: 379.59
Physical Form: Crystalline, Packaged under argon
Sensitivity: Air and Moisture Sensitive
Melting Point: 230-233°
UN No.: 3261
MDL No.: MFCD00001438
Formula:
C₁₀H₁₀Cl₂Hf MW: 379,58 g/mol Melting Pt: 230…233 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001438 CAS Number: 12116-66-4 EINECS: 235-177-5 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...