Bis(ethylcyclopentadienyl)hafnium dichloride ≥98%
Supplier: Thermo Fisher Scientific
H27592.03
H27592.06
H27592.03EA
0
EUR
H27592.03
H27592.06
Bis(ethylcyclopentadienyl)hafnium dichloride ≥98%
Bis(ethylcyclopentadienyl)hafnium dichloride
Formula:
C₁₄H₁₈Cl₂Hf MW: 435.69 g/mol |
MDL Number:
MFCD0014376 CAS Number: 78205-93-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...