2-Methyl-5-(methylthio)furan ≥99%

Supplier: Thermo Scientific
Warning

B21998.04 B21998.09
B21998.04EA 62.2 EUR
B21998.04 B21998.09
2-Methyl-5-(methylthio)furan ≥99%
2-Methyl-5-(methylthio)furan

Purity: 99% CAS No.: 13678-59-6 Molecular Formula: C7H14OS Formula Weight: 128.19 Flash Point: 50°(122°F) Odour: Stench Boiling Point: 79-81°/50mm UN No.: 1993 MDL No.: MFCD01208018

Formula: C₆H₈OS
MW: 128,19 g/mol
Boiling Pt: 79…81 °C (50 mmHg)
Density: 1,063
Flash Pt: 50 °C (122 °F)
Storage Temperature: Ambient
MDL Number: MFCD01208018
CAS Number: 13678-59-6
EINECS: 237-170-2
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR