DL-Ethionine 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-Amino-4-(ethylthio)butanoic acid
146170025
146170100
146170500
ACRO146170025EA
40.2
EUR
ACRO146170025
ACRO146170100
ACRO146170500
DL-Ethionine 98%
DL-Ethionine
Formula:
C₆H₁₃NO₂S MW: 163.24 g/mol Melting Pt: 265…268 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00063102 CAS Number: 67-21-0 EINECS: 200-647-0 |
Specification Test Results
Appearance | White to light yellow Crystalline powder or flakes |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥97.5 % |
GC | ≥94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...