N-Benzyloxycarbonyl-L-alaninol

Supplier: Thermo Fisher Scientific

Synonyms: Z-Ala-ol, (S)-Benzyl (1-hydroxypropan-2-yl)carbamate, (S)-(2-Hydroxy-1-methylethyl)carbamic acid benzyl ester, N-Cbz-L-Alaninol

H27066.03 H27066.06
H27066.03EA 63.8 EUR
H27066.03 H27066.06
N-Benzyloxycarbonyl-L-alaninol
N-Cbz-L-Alaninol

CAS No.: 66674-16-6 Molecular Formula: C11H15NO3 Formula Weight: 209.25 MDL No.: MFCD00672530

Formula: C₁₁H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD00672530
CAS Number: 66674-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR