Gold zinc alloy (95:5 w%) ≥99.99% (metals basis), slug, Length: 6.35mm (0.25in), Ø 6.35mm (0.25in)
Supplier: Thermo Scientific
43468.KG
43468.KJ
43468.KGEA
0
EUR
43468.KG
43468.KJ
Gold zinc alloy (95:5 w%) ≥99.99% (metals basis), slug, Length: 6.35mm (0.25in), Ø 6.35mm (0.25in)
Gold Zinc alloy (95:5 w%)
Purity: 99.99%
Molecular Formula: Au:Zn; 95:5 wt%
Physical Form: ≈3.7g/slug
MDL No.: MFCD02092916
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...