N-(2,3-Dimethylphenyl)-3-methylbenzamide ≥97%

Supplier: Thermo Scientific
Warning

H56395.MD H56395.03
H56395.MDEA 0 EUR
H56395.MD H56395.03
N-(2,3-Dimethylphenyl)-3-methylbenzamide ≥97%
N-(2,3-Dimethylphenyl)-3-methylbenzamide
Formula: C₁₆H₁₇NO
MW: 239,31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00443894
CAS Number: 292870-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR