Picolinic acid N-oxide 97%

Supplier: Thermo Fisher Scientific
Warning

131180050 131180250
ACRO131180050EA 35.3 EUR
ACRO131180050 ACRO131180250
Picolinic acid N-oxide 97%
Picolinic acid N-oxide
Formula: C₆H₅NO₃
Melting Pt: >150 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006198
CAS Number: 824-40-8
EINECS: 212-528-0

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point ≥145 °C
Water ≤0.5 % (K.F.)
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR