4-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 4-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester, 4-(4-Methylpiperazine-1-carbonyl)phenylboronic acid, pinacol ester, (4-Methylpiperazin-1-yl)(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanone

H28216.06 H28216.03
H28216.06EA 197 EUR
H28216.06 H28216.03
4-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester ≥97%
4-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester

Purity: 97% CAS No.: 832114-06-4 Molecular Formula: C18H27BN2O3 Formula Weight: 330.23 Melting Point: 110-113° MDL No.: MFCD05863914

Formula: C₁₈H₂₇BN₂O₃
MW: 330,23 g/mol
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD05863914
CAS Number: 832114-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR