2-Chloro-4-(4-piperidinyloxy)benzonitrile ≥98%
Supplier: Thermo Scientific
|
Danger
|
H50774.03
H50774.MD
H50774.03EA
283
EUR
H50774.03
H50774.MD
2-Chloro-4-(4-piperidinyloxy)benzonitrile ≥98%
2-Chloro-4-(4-piperidinyloxy)benzonitrile
Purity: 98%
CAS No.: 796600-10-7
Molecular Formula: C12H13ClN2O
Formula Weight: 236.70
UN No.: 3439
MDL No.: MFCD08060977
Formula:
C₁₂H₁₃ClN₂O MW: 236,70 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08060977 CAS Number: 796600-10-7 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...