3-(3-Piperidinyloxy)benzonitrile ≥99%
Supplier: Thermo Scientific
|
Warning
|
H50939.03
H50939.MD
H50939.03EA
336
EUR
H50939.03
H50939.MD
3-(3-Piperidinyloxy)benzonitrile ≥99%
3-(3-Piperidinyloxy)benzonitrile
Purity: 99%
CAS No.: 902836-93-5
Molecular Formula: C12H14N2O
Formula Weight: 202.26
MDL No.: MFCD08061079
Formula:
C₁₂H₁₄N₂O MW: 202,26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08061079 CAS Number: 902836-93-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...