(S)-(+)-3-Aminopiperidine dihydrochloride 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(S)-(+)-3-Piperidinamine dihydrochloride, (S)-Piperidin-3-amine dihydrochloride
H27700.03
H27700.03EA
132
EUR
H27700.03
(S)-(+)-3-Aminopiperidine dihydrochloride 98%
(S)-(+)-3-Aminopiperidine dihydrochloride
Purity: 98%
CAS No.: 334618-07-4
Molecular Formula: C5H12N2.2HCl
Formula Weight: 173.08
Melting Point: ca 210° dec.
Optical Rotation: +1.6° (c=1 in water)
Formula:
C₅H₁₄Cl₂N₂ MW: 173,08 g/mol Melting Pt: ∼ 210 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD03427036 CAS Number: 334618-07-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...