1-Boc-4-chloropiperidine 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
435010050
435010010
ACRO435010050EA
477
EUR
ACRO435010050
ACRO435010010
1-Boc-4-chloropiperidine 97%
1-Boc-4-chloropiperidine
Formula:
C₁₀H₁₈ClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD04115040 CAS Number: 154874-94-9 |
Specification Test Results
Appearance | Colorless to white Liquid or low melting solid |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...