(S)-1-Boc-2-ethylpiperazine ≥97%

Supplier: Thermo Fisher Scientific
Warning

H52738.MD H52738.03
H52738.MDEA 194 EUR
H52738.MD H52738.03
(S)-1-Boc-2-ethylpiperazine ≥97%
(S)-1-Boc-2-ethylpiperazine

Purity: 97% CAS No.: 325145-35-5 Molecular Formula: C11H22N2O2 Formula Weight: 214.30 MDL No.: MFCD07772100

Formula: C₁₁H₂₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD07772100
CAS Number: 325145-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR