Benzyltrimethylammonium tribromide ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Mono(N,N,N-trimethyl-1-phenylmethanaminium) tribromide, Benzyl trimethyl ammonium tribromide
B24470.06
B24470.14
B24470.06EA
40.3
EUR
B24470.06
B24470.14
Benzyltrimethylammonium tribromide ≥98%
Benzyltrimethylammonium tribromide
Purity: 98%
CAS No.: 111865-47-5
Molecular Formula: C6H5CH2N(CH3)3Br3
Formula Weight: 389.97
Melting Point: 99-101°
MDL No.: MFCD00134423
Formula:
C₁₀H₁₆Br₃N Melting Pt: 99…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134423 CAS Number: 111865-47-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...