2-(Methylamino)-5-pyrimidineboronic acid pinacol ester 96%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Methylaminopyrimidine-5-boronic acid, pinacol ester, N-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
H54682.MD
H54682.03
H54682.MDEA
98.5
EUR
H54682.MD
H54682.03
2-(Methylamino)-5-pyrimidineboronic acid pinacol ester 96%
2-(Methylamino)-5-pyrimidineboronic acid pinacol ester
Purity: 96%
CAS No.: 904326-88-1
Molecular Formula: C11H18BN3O2
Formula Weight: 235.09
Storage Temperature: 2°C to 8°C
Melting Point: 122-125°
MDL No.: MFCD11878345
Formula:
C₁₁H₁₈BN₃O₂ MW: 235,09 g/mol Melting Pt: 122…125 °C |
MDL Number:
MFCD11878345 CAS Number: 904326-88-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...