Diethoxymethylphenylsilane ≥97%

Supplier: Thermo Scientific
Warning

B23684.09 B23684.18
B23684.09EA 25.9 EUR
B23684.09 B23684.18
Diethoxymethylphenylsilane ≥97%
Diethoxymethylphenylsilane

Purity: 97+% CAS No.: 775-56-4 Molecular Formula: (CH3CH2O)2Si(CH3)C6H5 Formula Weight: 210.35 Flash Point: 88°(190°F) Refractive Index: 1.4696 Boiling Point: 117-118°/30mm Beilstein Reference: 2938396 MDL No.: MFCD00048541

Formula: C₁₁H₁₈O₂Si
Boiling Pt: 117…118 °C (30 mmHg)
Density: 0,963
Flash Pt: 88 °C (190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00048541
CAS Number: 775-56-4
EINECS: 212-275-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR